Learn More
Cytek Biosciences Inc Gt amouse IgG Anti Secondary
Supplier: Cytek Biosciences Inc 308054U500
Product Name: Biotin Goat Anti-Mouse IgG (H&L)
Product Class: Antibody
Type: Secondary
Product Name: Biotin Goat Anti-Mouse IgG (H&L)
Cytek Catalog Number: 30-8054-U500
Marker: IgG (H&L)
Target Species: Mouse
Reported Cross Reactivity: Mouse
Clonality: Polyclonal
Host Species: Goat
Conjugation: Biotin
Size: 500 μg
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.