Learn More
Cytek Biosciences Inc Gt aMu IgG Fc HRP Anti
Supplier: Cytek Biosciences Inc 728093M001
Product Name: HRP Goat Anti-Mouse IgG Fc
Product Class: Antibody
Type: Secondary
Product Name: HRP Goat Anti-Mouse IgG Fc
Cytek Catalog Number: 72-8093-M001
Marker: IgG Fc
Target Species: Mouse
Reported Cross Reactivity: Mouse
Clonality: Polyclonal
Host Species: Goat
Conjugation: HRP
Size: 1 mg
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.