Learn More
Cytek Biosciences Inc Gt aMu IgG2b PE Anti
Supplier: Cytek Biosciences Inc 508035U100
Product Name: PE Goat Anti-Mouse IgG2b
Product Class: Antibody
Type: Secondary
Product Name: PE Goat Anti-Mouse IgG2b
Cytek Catalog Number: 50-8035-U100
Marker: IgG2b
Target Species: Mouse
Reported Cross Reactivity: Mouse
Clonality: Polyclonal
Host Species: Goat
Conjugation: PE
Size: 100 μg
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.