Learn More
Cytek Biosciences Inc Gt aMu IgM Anti Secondary
Supplier: Cytek Biosciences Inc 708064M002
Product Name: Unconjugated Goat Anti-Mouse IgM (H&L)
Product Class: Antibody
Type: Secondary
Product Name: Unconjugated Goat Anti-Mouse IgM (H&L)
Cytek Catalog Number: 70-8064-M002
Marker: IgM (H&L)
Target Species: Mouse
Reported Cross Reactivity: Mouse
Clonality: Polyclonal
Host Species: Goat
Conjugation: Unconjugated
Size: 2 mg
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.