Learn More
Immunology Consultants anti-Mouse IgG h+l Antibody (Goat) - FITC Conjugated
SDP
Supplier:  Immunology Consultants GGHL90F
Encompass_Preferred
This FITC‑conjugated goat antibody provides direct fluorescent detection of mouse IgG, supporting flow cytometry, immunofluorescence microscopy, and fluorescence‑based ELISA formats. The conjugate delivers bright, stable signal with low nonspecific binding, enabling clear visualization of mouse primary antibodies in cell‑based and fixed‑tissue applications. GGHL‑90F is ideal for fluorescence workflows requiring consistent, high‑clarity detection.
Catalog No. 50-196-2196