Learn More
Jackson Immuno Research Labs R-Phycoerythrin AffiniPure F(ab')₂ Fragment Goat Anti-Mouse IgG (H+L), 1ml
SDP
Supplier:  Jackson Immuno Research Labs 115116146
Encompass_Preferred
R-Phycoerythrin-AffiniPure F(ab')2 Fragment Goat Anti-Mouse IgG (H+L) (min X Hu,Bov,Hrs,Rb,Sw Sr Prot)
Catalog No. NC9696881