Learn More
Cytek Biosciences Inc Gt aRb IgG Anti Secondary
Supplier: Cytek Biosciences Inc 708067M002
Product Name: Unconjugated Goat Anti-Rabbit IgG (H&L)
Product Class: Antibody
Type: Secondary
Product Name: Unconjugated Goat Anti-Rabbit IgG (H&L)
Cytek Catalog Number: 70-8067-M002
Marker: IgG (H&L)
Target Species: Rabbit
Reported Cross Reactivity: Rabbit
Clonality: Polyclonal
Host Species: Goat
Conjugation: Unconjugated
Size: 2 mg
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.