Learn More
Cytek Biosciences Inc Gt aRb IgG Fc HRP Anti
Supplier: Cytek Biosciences Inc 728110M001
Product Name: HRP Goat Anti-Rabbit IgG Fc
Product Class: Antibody
Type: Secondary
Product Name: HRP Goat Anti-Rabbit IgG Fc
Cytek Catalog Number: 72-8110-M001
Marker: IgG Fc
Target Species: Rabbit
Reported Cross Reactivity: Rabbit
Clonality: Polyclonal
Host Species: Goat
Conjugation: HRP
Size: 1 mg
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.