Learn More
Biocare Medical, LLC Goat-on-Rodent HRP-Polymer, 25 ml
SDP
Supplier:  Biocare Medical, LLC GHP516H
Encompass_Preferred
Goat-on-Rodent HRP-Polymer is for goat primary antibodies on mouse, rat or human tissues and labeled with horseradish peroxidase. Goat antibodies are advantageous as the detection has minimum cross reactivity to mouse, rat or human IgG.
Catalog No. 50-828-63