Learn More
Waters Corp GTxResolve Slalom Column, 250 Å, 2.5 μm, 4.6 × 300 mm
Supplier:  Waters Corp 176006046
Encompass_Preferred
This column is designed for high-resolution slalom chromatography, effectively separating large nucleic acids and plasmids. It ensures robust, reproducible stability and consistent performance for fast, cost-effective assessment of dsRNA and plasmid quality.
  • Provides high resolution and precision separation of large nucleic acids and plasmids.
  • Ensures robust and reproducible stability and consistent performance.
  • Incorporates MaxPeak High Performance Surfaces (HPS) for minimized analyte/surface interactions.
  • Increases analyte recovery, sensitivity, and reproducibility.
  • Reduces column conditioning and passivation times, saving development time.
  • Improves sensitivity, peak shapes, and recovery of metal-sensitive analytes.
Catalog No. NC3924973