Learn More
Medchemexpress LLC Guadecitabine sodium | 929904-85-8 | C18H23N9NaO10P | 1 ML
SDP
Supplier:  Medchemexpress LLC HY1522910MM/1ML
Encompass_Preferred
Guadecitabine sodium is a second-generation DNA methyltransferases (DNMT) inhibitor for research of acute myeloid leukemia (AML) and myelodysplastic syndromes (MDS). It is for research use only and not sold to patients.
  • Second-generation DNA methyltransferases (DNMT) inhibitor
  • Induces dose-dependent increase in p16 expression
  • Inhibits DNA methylation
  • Effective in reducing DNA methylation and retarding tumor growth in vivo
  • Reactivates p16 gene expression
  • Better tolerated in vivo than 5-Aza-CdR
  • Involved in numerous clinical trials for various conditions
Catalog No. 50-003-73872