Learn More
Medchemexpress LLC Guadecitabine sodium | 929904-85-8 | C18H23N9NaO10P | 100 MG
SDP
Supplier:  Medchemexpress LLC HY15229100MG
Encompass_Preferred
Guadecitabine sodium (SGI-110 sodium) is a second-generation DNA methyltransferases (DNMT) inhibitor. It is used in research for acute myeloid leukemia (AML) and myelodysplastic syndromes (MDS).
  • Inhibits DNA methylation at the 5′-region
  • Induces p16 gene expression
  • Reduces DNA methylation in vivo
  • Retards tumor growth in vivo
  • Reactivates p16 gene expression in vivo
  • Better tolerated than 5-Aza-CdR in vivo
Catalog No. 50-003-73545