Learn More
Cosmo Bio Usa Inc Guaiacol Detection Kit 100 Tests
SDP
Supplier:  Cosmo Bio Usa Inc KYO08921
Encompass
Detection of guaiacole Alicyclobacillus acidoterrestris is a gram-positive, spore-forming bacillus that occurs widely in nature. This species have been studied substantially after the incident of turbidity of transparent apple juice in 1984. The Alicyclobacillus genus bacteria, which peculiarly grow under relatively high temperature and acid conditions, are called Thermo-acidophilic Bacilli (TAB) ##(1, 2, 3)###.
Catalog No. NC9002540