Learn More
Medchemexpress LLC Guaiazulene | 489-84-9 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYN6951R10MG
Encompass_Preferred
Guaiazulene (Standard) is an analytical standard used for research and analytical applications. This bicyclic sesquiterpene can cross the blood-brain barrier and exhibits various biological activities, including anti-inflammatory, antioxidant, hepatoprotective, antibacterial, and anti-tumor properties. It is also commonly used as a colorant in cosmetics and shows in vitro cytotoxicity to rat neuronal cells and N2a neuroblastoma cells at high concentrations.
  • Anti-inflammatory properties
  • Antioxidant properties
  • Hepatoprotective properties
  • Antibacterial properties
  • Anti-tumor properties
  • Used as a colorant in cosmetics
  • Utilized in qualitative, quantitative, and methodological research experiments (HPLC, GC, MS)
Catalog No. 50-004-19882