Learn More
Medchemexpress LLC Guaiazulene (Standard) | 489-84-9 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYN6951R100MG
Encompass_Preferred
Guaiazulene (Standard) is an analytical standard intended for research and analytical applications. This bicyclic sesquiterpene can cross the blood-brain barrier and exhibits various biological activities including anti-inflammatory, antioxidant, hepatoprotective, antibacterial, and anti-tumor properties. It is also used as a colorant in cosmetics.
  • Analytical standard for research and applications
  • Can cross the blood-brain barrier
  • Exhibits anti-inflammatory properties
  • Exhibits antioxidant properties
  • Exhibits hepatoprotective properties
  • Exhibits antibacterial properties
  • Exhibits anti-tumor properties
  • Commonly used as a colorant in cosmetics
  • Used as a reference standard for assays
  • Frequently used in qualitative, quantitative, and methodological research experiments involving HPLC, GC, and MS
Catalog No. 50-004-18337