Learn More
Apexbio Technology LLC Guaifenesin 93-14-1 5g
SDP
Supplier:  Apexbio Technology LLC A84425000
Encompass_Preferred
Guaifenesin (93-14-1) is a small-molecule expectorant designed to modulate respiratory epithelial cell secretion thereby regulating mucus viscosity and bronchial mucus clearance in the respiratory tract Guaifenesin exerts its biological activity primarily through enhancement of respiratory tract hydration and reduction of mucus viscosity via direct modulation of respiratory epithelial cells Based on these pharmacological properties guaifenesin holds research potential in studies of mucus clearance mechanisms bronchial secretion regulation and potential application in neuromuscular activity modulation with exploratory investigations in fibromyalgia symptom management and reproductive biology
Catalog No. 50-000-95965