Learn More
Medchemexpress LLC Guaifenesin | 93-14-1 | 99.9% | 1 G
SDP
Supplier:  Medchemexpress LLC HYB02641G
Encompass_Preferred
Guaifenesin, also known as Guaiacol glyceryl ether, is an expectorant derived from the guaiac resin of Guajacum officinale Linné. It is used to alleviate cough discomfort by increasing sputum volume and decreasing its viscosity, thereby promoting effective cough.
  • Alleviates cough discomfort
  • Increases sputum volume
  • Decreases sputum viscosity
  • Promotes effective cough
  • Inhibits mucin release in epithelial cultures (in vitro)
  • Inhibits IL-13-induced MUC5AC content and secretion in primary human airway epithelial cells (in vitro)
Catalog No. 50-004-06914