Learn More
Medchemexpress LLC Guaifenesin (guaiacol glyceryl ether) | 93-14-1 | 99.7% | 10 MG
SDP
Supplier:  Medchemexpress LLC HYB0264R10MG
Encompass_Preferred
Guaifenesin (standard) is an analytical reference standard of guaifenesin intended for research and analytical applications, including assay, method development, and quality control. The material is provided as a high-purity solid suitable for chromatographic and mass spectrometric analysis.
  • Analytical reference standard for assay and method development.
  • High purity: 99.65%.
  • Supplied in accurately weighed small quantities suitable for standard preparation (e.g., 10 MG).
  • Suitable for chromatographic and spectrometric analyses.
  • Identifying information included for analysis: CAS number 93-14-1 and molecular weight 198.22 g/mol.
Catalog No. 50-002-96869