Learn More
Medchemexpress LLC HY-B0566 100mg Medchemexpress, Guanabenz (Acetate) CAS:23256-50-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB0566100MG
Encompass_Preferred
Medchemexpress, HY-B0566 100mg Guanabenz (Acetate) CAS:23256-50-0 Guanabenz (Acetate) (BR-750) is an alpha-2 selective adrenergic agonist used as an antihypertensive agent. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2602