Learn More
Selleck Chemical LLC Guanabenz Acetate S4065-10mM/1mL
SDP
Supplier:  Selleck Chemical LLC S406510MM/1ML
Encompass_Preferred
Guanabenz Acetate (WY-8678) is an selective agonist of 2a-adrenergic receptor 2b-adrenergic receptor and 2c-adrenergic receptor with pEC50 of 8 25 7 01 and 5 respectively
Catalog No. 50-310-9684