Learn More
Selleck Chemical LLC Guanabenz Acetate 50mg 23256-50-0 WY-8678 Acetate
SDP
Supplier:  Selleck Chemical LLC S406550MG
Encompass_Preferred
Guanabenz Acetate (WY-8678) is an selective agonist of ?2a-adrenergic receptor, ?2b-adrenergic receptor and ?2c-adrenergic receptor with pEC50 of 8.25, 7.01 and ~5, respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-3624