Learn More
Apexbio Technology LLC Guanethidine (sulfate) 645-43-2 5g
SDP
Supplier:  Apexbio Technology LLC B86835000
Encompass_Preferred
Guanethidine sulfate (CAS 645-43-2) is a small-molecule inhibitor targeting peripheral adrenergic neurons It is designed to selectively inhibit the secretion of norepinephrine from sympathetic nerve endings thereby modulating sympathetic neurotransmission in cardiovascular pathways Guanethidine sulfate exerts its biological activity primarily through neuronal uptake via amine transporters and interference with norepinephrine release Based on these pharmacological properties Guanethidine sulfate holds research potential in the study of autonomic regulation of blood pressure neurotransmitter dynamics and mechanisms underlying sympathetic nervous system activity
Catalog No. 50-000-89331