Learn More
Apexbio Technology LLC Guanfacine 29110-47-2 50mg
SDP
Supplier:  Apexbio Technology LLC B339450
Encompass_Preferred
Guanfacine (CAS 29110-47-2) is an orally active highly selective agonist of the 2A-adrenergic receptor By stimulating 2A receptors in the prefrontal cortex guanfacine modulates noradrenergic signaling pathways resulting in decreased blood pressure and sedative effects In biomedical research guanfacine is widely utilized to investigate the role of the noradrenergic system in cognitive function particularly in models exhibiting prefrontal cortex dysfunction It is frequently applied in studies on attention deficit hyperactivity disorder (ADHD) Tourette syndrome and related neuropsychiatric conditions to explore underlying mechanisms and evaluate potential therapeutic strategies
Catalog No. 50-000-94247