Learn More
Medchemexpress LLC Guanidine-d5 (hydrochloride) | 108694-93-5 | MFCD00144417 | 98.0% | 101.57 g/mol | CD6ClN3 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYB0178AS110MG
Encompass_Preferred
Guanidine-d5 hydrochloride is the deuterium-labeled analogue of guanidine hydrochloride supplied as a solid research reagent and analytical standard. It is used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, and LC-MS. Certificate of analysis and safety data sheet are provided by the manufacturer.
  • Deuterium-labeled guanidine hydrochloride suitable for analytical workflows.
  • High isotopic enrichment and 98.0% purity for reliable quantification.
  • Useful as an internal standard for NMR, GC-MS, and LC-MS methods.
  • Solid form available in small quantities (10 mg) for analytical use.
Catalog No. 50-002-98862