Learn More
Medchemexpress LLC Guanidine hydrochloride | 50-01-1 | 98.0% | 500 G
SDP
Supplier:  Medchemexpress LLC HYB0178A500G
Encompass_Preferred
Guanidine hydrochloride (Guanidinium chloride) is a strong chaotrope and a potent denaturant of proteins, intended strictly for research purposes.
  • Facilitates the refolding of acid-unfolded apomyoglobin and cytochrome c at low concentrations.
  • Induces co-operative unfolding of the molten globule state at concentrations exceeding 1 M.
  • Does not interfere with disulfide bonds, preserving ordered protein structure.
  • Effective in causing the loss of the stable [PSI+] element from yeast cells at millimolar concentrations.
  • A 5 mM concentration in growth media cures [PSI+] and other yeast prions.
  • Markedly diminishes Hsp104-mediated basal and acquired thermotolerance.
  • Impairs Hsp104's capacity to restore activity in thermally denatured luciferase.
  • Significantly lowered the death rate of infant mice infected with coxsackievirus A16 in studies.
Catalog No. 50-004-07252