Learn More
Medchemexpress LLC Guanidinoacetic acid-d2 | 1173020-63-7 | MFCD04118279 | 99.9% | 119.12 | C3H5D2N3O2 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYW021448S10MG
Encompass_Preferred
Glycocyamine-d2 (guanidinoacetic acid-d2) is a deuterium-labeled stable isotope of glycocyamine used as an internal standard and tracer in analytical and biochemical research. Supplied as a white to off-white solid with high chemical purity and defined isotopic labeling, it supports reliable quantitation and metabolic studies.
  • Deuterium-labeled internal standard for mass spectrometry and LC-MS assays.
  • High chemical purity for accurate quantitation.
  • Defined isotopic labeling for metabolic tracing and flux analysis.
  • Solid form simplifies handling and storage.
  • Stable under recommended storage to preserve isotopic integrity.
Catalog No. 50-003-00762