Learn More
Medchemexpress LLC HY-Y1055 100mg Medchemexpress, Guanine CAS:73-40-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYY1055100MG
Encompass_Preferred
Medchemexpress, HY-Y1055 100mg Guanine CAS:73-40-5 Guanine is one of the fundamental components of nucleic acids (DNA and RNA). Guanine is a purine derivative, consisting of a fused pyrimidine-imidazole ring system with conjugated double bonds. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0770