Learn More
Abcam Guanosine 3',5'-cyclic monophosphate sodium salt (cGMP), cyclic nucleotide second messenger, 250MG
Supplier:  Abcam AB120805250MG
Encompass_Preferred
MW 367.19 Da, Purity >99%. Cyclic nucleotide second messenger derived from guanosine triphosphate. Generated by guanylate cyclases. Involved in a number of cellular processes; downstream targets include cyclic nucleotide gated channels, cGMP-regulated phosphodiesterases and cGMP-dependent kinases.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7249