Learn More
Medchemexpress LLC Guanosine-5'-triphosphate disodium salt | 56001-37-7 | 99.6% | 567.14 | 1 G
SDP
Supplier:  Medchemexpress LLC HYW0107371G
Encompass_Preferred
Guanosine 5'-triphosphate (5'-GTP) trisodium salt is a G protein signaling activator and a high-energy precursor in the biosynthesis of nucleotide units in DNA and RNA. It can promote myogenic cell differentiation by upregulating miRNA (miR133a, miR133b) and myogenic regulatory factor expression, and by inducing human myogenic precursor cells to release exosomes containing guanosine molecules. It holds promise for research in biosynthesis and skeletal muscle regeneration.
  • G protein signaling activator
  • High-energy precursor in nucleotide biosynthesis for DNA and RNA
  • Promotes myogenic cell differentiation
  • Upregulates miRNA (miR133a, miR133b) and myogenic regulatory factors
  • Induces human myogenic precursor cells to release exosomes containing guanosine molecules
Catalog No. 50-003-66321