Learn More
Medchemexpress LLC Guanoxabenz hydrochloride | 23256-40-8 | 283.54 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYU00123A10MG
Encompass_Preferred
Guanoxabenz hydrochloride is an α2 adrenergic receptor agonist, with a Kᵢ of 4000 nM and a fully activated form having a 40 nM affinity for an α2A adrenoceptor. It functions as a centrally active antihypertensive drug, demonstrated to reduce locomotor activity and influence EEG in animal models. Its high-affinity binding is influenced by N-hydroxyguanidine analogs and metabolic inhibitors.
  • Functions as an α2 adrenergic receptor agonist.
  • Acts as a centrally active antihypertensive drug.
  • High-affinity binding inhibited by N-hydroxyguanidine analogs.
  • Spleen cytosolic fraction converts Guanoxabenz to guanabenz.
  • Reduces locomotor activity in rats dose-dependently.
  • Influences EEG and behavioral effects.
Catalog No. 50-004-47711