Learn More
Medchemexpress LLC Guanoxabenz hydrochloride | 23256-40-8 | 283.54 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYU00123A100MG
Encompass_Preferred
Guanoxabenz hydrochloride (Hydroxyguanabenz) is an α2 adrenergic receptor agonist with a Ki of 4000 nM and a fully activated form of 40 nM for an α2A adrenoceptor.
  • Guanoxabenz is known as a centrally active antihypertensive drug.
  • High affinity Guanoxabenz binding is induced in rat brain membranes after the addition of NADH or NADPH cofactors.
  • The rat cerebral cortex contains an enzymatic activity that may activate Guanoxabenz, leading to the formation of a metabolite with high affinity for alpha 2-adrenoceptors.
  • Guanoxabenz (0.1-3 mg/kg, i.p.) causes a dose-related reduction in locomotor activity.
  • A dose of 1 mg/kg induces pronounced and sustained behavioral hypoactivity in rats.
Catalog No. 50-004-49616