Learn More
Medchemexpress LLC Guanoxabenz hydrochloride | 23256-40-8 | 283.54 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYU00123A
Encompass_Preferred
Guanoxabenz hydrochloride, also known as Hydroxyguanabenz hydrochloride, is an α2 adrenergic receptor agonist. It acts as a centrally active antihypertensive drug and is intended for research use only.
  • Acts as an α2 adrenergic receptor agonist.
  • Displays a Ki of 4000 nM for an α2A adrenoceptor, with 40 nM for its fully activated form.
  • Recognized as a centrally active antihypertensive drug.
  • High-affinity binding can be inhibited by N-hydroxyguanidine analogs and various metabolic inhibitors.
  • Reduction to guanabenz, with higher affinity for alpha2A-adrenoceptors, is mediated by spleen cytosolic fraction.
  • Induces high-affinity binding in rat brain membranes with NADH or NADPH cofactors.
  • Causes a dose-related reduction in locomotor activity in vivo.
Catalog No. 50-004-48013