Learn More
Medchemexpress LLC Guanoxabenz hydrochloride | 23256-40-8 | 283.54 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYU00123A50MG
Encompass_Preferred
Guanoxabenz hydrochloride is an α2 adrenergic receptor agonist. It has a Ki of 4000 nM and the fully activated form has a Ki of 40 nM for an α2A adrenoceptor.
  • Acts as an α2 adrenergic receptor agonist
  • Features a Ki of 4000 nM
  • Fully activated form has a Ki of 40 nM for an α2A adrenoceptor
Catalog No. 50-004-48949