Learn More
Medchemexpress LLC Guanylyl Cyclase alpha 1 antibody (YA2937) | 10 UL
SDP
Supplier:  Medchemexpress LLC HYP8319210UL
Encompass_Preferred
Guanylyl Cyclase alpha 1 Antibody (YA2937) is a recombinant, monoclonal, Rabbit-derived IgG antibody designed for research use. This non-conjugated antibody targets Guanylyl Cyclase alpha 1 and is supplied as a liquid formulation.
  • Targets Guanylyl Cyclase alpha 1
  • Rabbit-derived IgG monoclonal antibody
  • Affinity purified
  • Non-conjugated
  • Suitable for Western Blot (WB) applications
  • Reacts with human samples
  • Subcellular localization in cytoplasm
Catalog No. 50-004-43192