Learn More
Mac-Mod Avantor Alltima C18, 100 Å, 5 µm, 2.1 x 7.5 mm Guard Cartridges (10/pk)
SDP
Supplier:  Mac-Mod 5174010/N / 5174010/N
Encompass
vantor Alltima columns combines a range of different phase chemistries with base deactivated silica. Avantor Alltima columns provides stable bonding for long column lifetimes. Avantor Alltima columns are made of high purity silica that provide excellent column stability with multiple selectivity options.
Catalog No. 50-001-43208