Learn More
Medchemexpress LLC GUCY2C/Guanylyl cycl 20ug

Supplier: Medchemexpress LLC HYP7166620UG
Recombinant rat Guanylyl cyclase C (GUCY2C) protein expressed in HEK293 cells and supplied with Myc and His affinity tags. The protein is provided in small microgram pack sizes for biochemical, receptor-binding, and functional studies of GC-C signaling. Sequence range and purity are suitable for assay development and antibody production.
- Recombinant rat GUCY2C expressed in mammalian HEK293 cells.
- Includes Myc and His affinity tags for detection and purification.
- Suitable for receptor binding and signaling assays.
- Supplied in low microgram pack sizes for small-scale experiments.
- Sequence range and purity appropriate for biochemical studies.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.