Learn More
Selleck Chemical LLC Guggulsterone E&Z 5mg 95975-55-6
SDP
Supplier:  Selleck Chemical LLC S37925MG
Encompass_Preferred
Guggulsterone is one of the active constituent of Commiphora mukul. It occurs in two isomeric forms, namely Z-GS and E-GS. Guggulsterone act as antagonist ligands for the bile acid receptor, farnesoid X receptor, and as active ingredients responsible for the hypolipidemic activity. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8347