Learn More
Grainger Guide Bar For Hydraulic Power Saw L 15in
Supplier:  Grainger 16V976
Encompass_Preferred
The Grainger 16V976 is a Dayton 1/3 HP, 115/230V AC, 1725 RPM, 56C Frame, Open Drip Proof, Ball Bearing, Electric Motor. It has a 1/3 horsepower, operates on 115/230 volt AC power, has a 1725 RPM speed, uses a 56C frame, is open drip proof, contains ball bearings, and is designed for general purpose applications. The motor is thermally protected, reversible, and suitable for OEM and MRO applications.
Catalog No. 50-276-7735