Learn More
Med Vet International Guide for Long / Neutral 2.4mm Screw, J1073LNG
Supplier: Med Vet International JOR1073LNG
To exert compression at a fracture site, the pilot bit must be positioned precisely in the slot in the compression plate. The load/neutral drill guide has two sleeves, one for compression (load) and one where no compression is exerted (neutral). There is one combined guide for each screw size.Load/Neutral Drill Guide for 2.4mm plates. Uses 1.8mm drill bit.