Learn More
Rockland Immunochemicals GUINEA PIG COMPLEMENT (fresh frozen) - C300-0010
SDP
Supplier:  Rockland Immunochemicals C3000010
Encompass
Special processing techniques are used to yield products with high complement activity and low background cytotoxicity. Guinea Pig Complement is suitable for CFT and SRH.
Catalog No. NC1463557