Learn More
Kingfisher Biotech Inc Ovine TNF alpha ELISA - Less than or equal to 20 plates (96-well)
SDP
Supplier:  Kingfisher Biotech Inc DIY1463GP003
Encompass_Preferred
The Ovine TNF alpha (TNFSF2) ELISA contains capture antibody, standard, and detection antibody for development of a Ovine TNF alpha ELISA. The antibodies have been determined to function in an ELISA with the standard provided. Optimal buffers, concentrations, incubation times, incubation temperatures, and methods for the ELISA have not been determined. This allows for complete flexibility to provide optimal dynamic range and sensitivity based on your sample type. Alternate names include TNF, DIF, TNF-alpha, TNFA, TNFSF2, Tumour necrosis factor, tumor necrosis factor, TNLG1F, Tumor necrosis factor alpha. The quantities of components provided are not matched. Components may also be purchased separately. For research use only. Made in the USA
Catalog No. 50-228-5475