Learn More
American Research Products Inc Guinea pig TGFb1 ELISA Kit
Supplier:  American Research Products Inc RDRTGFB1GU
Encompass
This Guinea pig Transforming Growth Factor Beta 1 (TGFb1) ELISA Kit is a precoated Sandwich ELISA assay designed to detect and quantify the target analyte in serum platelet-poor plasma tissue homogenates cell culture supernatants and other biological fluids This product is for Research Use Only
Catalog No. 50-004-56497