Learn More
Cayman Chemical GunIn-2-deoxyL-rIbofura 2.5mg
SDP
Supplier:  Cayman Chemical 384652.5 MG
Encompass_Preferred
An isomer of dGTP and a telomerase inhibitor (Ki = 15 µM for the human enzyme)
Catalog No. 50-291-7115