Learn More
Cayman Chemical Gunylyl ImIdodIphosphATlI 10mg
SDP
Supplier:  Cayman Chemical 1688010 MG
Encompass_Preferred
A nonhydrolyzable GTP analog that binds to and irreversibly activates G proteins; potently stimulates adenylate cyclase
Catalog No. 50-296-7157