Learn More
Medchemexpress LLC Guretolimod (hydrochloride) | 98.7% | 550.01 | 100 MG

Supplier: Medchemexpress LLC HY139575A100MG
Guretolimod hydrochloride is a Toll-like receptor 7 (TLR7) agonist, suitable for research applications. It is supplied as an off-white to light yellow solid.
- Targets toll-like receptor 7 (TLR7)
- Appears as an off-white to light yellow solid
- Recommended storage at -20°C, sealed, away from moisture and light
- Soluble in DMSO at 100 mg/mL (181.81 mM)
- For research use only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.