Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Guretolimod hydrochloride | 98.7% | 550.01 | 50 MG
SDP

Supplier:  Medchemexpress LLC HY139575A50MG

Encompass_Preferred

Guretolimod hydrochloride is a Toll-like receptor 7 (TLR7) agonist for research use only, not intended for patient use. It is a solid with an off-white to light yellow color.

  • Acts as a Toll-like receptor 7 (TLR7) agonist
  • For research use only
  • Purity of 98.7%
  • Molecular weight of 550.01
  • Solid appearance, off-white to light yellow color
  • Soluble in DMSO at 100 mg/mL
  • Store at -20°C, sealed, away from moisture and light; in solvent at -80°C for 6 months or -20°C for 1 month

Catalog No. 50-003-49774


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.