Learn More
Carbosynth LLC. Gurmarin, 1mg, Disulfide bonds: Cys3-Cys18, Cys10-Cys23, and Cys17-Cys33, MW: (M.W. 4208.95), Molecular formula: C187H276N46O53S6, CAS# [138464-10-5]
SDP
Supplier:  Carbosynth LLC. GUR3810PI1MG
Encompass_Preferred
Synonyms: Pyr-Gln-Cys-Val-Lys-Lys-Asp-Glu-Leu-Cys-Ile-Pro-Tyr-Tyr-Leu-Asp-Cys-Cys-Glu-Pro-Leu-Glu-Cys-Lys-Lys-Val-Asn-Trp-Trp-Asp-His-Lys-Cys-Ile-Gly-OH, Application: A 35 amino acid peptide, originally from the leaves of the Gymnema sylvetre vine, known to suppress sweet taste and a potential therapeutic treatment of type II diabetes, Storage Temperature: -20°C, References: J.I. Fletcher, et al., Eur. J. Biochem., 264, 525 (1999)., M. Sigoillot, et al., Appl. Microbiol. Biotechnol., 96, 619 (2012)., Y. Ninomiya, et al., Am J Physiol., 274, R1324 (1998).
Catalog No. 50-168-5788