Learn More
Medchemexpress LLC HY-103018 25mg Medchemexpress, Gusacitinib CAS:1425381-60-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10301825MG
Encompass_Preferred
Medchemexpress, HY-103018 25mg Gusacitinib CAS:1425381-60-7 Gusacitinib (ASN-002) is an orally active and potent dual inhibitor of spleen tyrosine kinase (SYK) and janus kinase (JAK) with IC50 values of 5-46 nM. Gusacitinib has anti-cancer activity in both solid and hematological tumor types. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0655