Learn More
Medchemexpress LLC HY-100809 10mg Medchemexpress, Guvacine hydrochloride CAS:6027-91-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10080910MG
Encompass_Preferred
Medchemexpress, HY-100809 10mg Guvacine hydrochloride CAS:6027-91-4 Guvacine hydrochloride is an alkaloid from the nut of Areca catechu, acts as an inhibitor of GABA transporter , and dispalys modest selectivity for cloned GABA transporters with IC 50 s of 14 μM (human GAT-1), 39 μM (rat GAT-1), 58 μM (rat GAT-2), 119 μM (human GAT-3), 378 μM (rat GAT-3), and 1870 μM (human BGT-3). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1957