Learn More
Medchemexpress LLC Guvacine hydrobromide | 6027-92-5 | 98.0% | 208.05 | C6H10BrNO2 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYN2482A10MG
Encompass_Preferred
Guvacine hydrobromide is the hydrobromide salt of guvacine, a GABA uptake inhibitor used in neuroscience research to probe GABA transporter function and inhibitory neurotransmission. It is supplied as a solid salt form with high purity suitable for biochemical and pharmacological studies.
  • Potent GABA uptake inhibitor active at GAT transporters.
  • Hydrobromide salt for improved water solubility compared with the free base.
  • High purity (~98.0%) appropriate for in vitro assays.
  • Supplied as a stable solid for routine storage and handling.
  • Useful as a research tool in studies of inhibitory neurotransmission and transporter pharmacology.
Catalog No. 50-002-89656